Learn More
2-((6-Methoxy-3-methyl-2,4-dioxo-3,4-dihydropyrimidine-1(2H)-yl)methyl)benzonitrile, TRC
Product Code. 30474512
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30474512 100 mg 100mg
30470326 50 mg 50mg
30476436 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30474512 Supplier Toronto Research Chemicals Supplier No. TRCM330608100MG
 Restricted Item
Forventet forsendelse: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2514763-78-9
Molecular Formula C14H13N3O3
Molecular Weight (g/mol) 271.27
Synonym Alogliptin impurity
IUPAC Name 2-[(6-methoxy-3-methyl-2,4-dioxopyrimidin-1-yl)methyl]benzonitrile
SMILES COC1=CC(=O)N(C)C(=O)N1Cc2ccccc2C#N
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C14H13N3O3/c1-16-12(18)7-13(20-2)17(14(16)19)9-11-6-4-3-5-10(11)8-15/h3-7H,9H2,1-2H3
Formula Weight 271.0957
Chemical Name or Material 2-((6-Methoxy-3-methyl-2,4-dioxo-3,4-dihydropyrimidine-1(2H)-yl)methyl)benzonitrile