Få mere at vide
2,6-Dimethylaniline-d6, TRC
Artikelnummer. 30374261
Skift visning
Mængde:
2.5 mg
Pakningsstørrelse:
2.5mg
Artikelnummer. Mængde unitSize
30374261 2.5 mg 2.5mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30374261 Leverandør Toronto Research Chemicals Leverandørnr. TRCD4621972.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2,6-Dimethylaniline-d6 (o-Xylidine-d6)
CAS 919785-81-2
Renhedsgrad noter HPLC
Molekylær formel C8 2H6 H5 N
Mængde 2.5 mg
Synonym 2,6-Di(methyl-d3)benzenamine (ACI), 2,6-Bis(trideuteriomethyl)aniline,2,6-Dimethylaniline D6 (dimethyl D6)
InChI formel InChI=1S/C8H11N/c1-6-4-3-5-7(2)8(6)9/h3-5H,9H2,1-2H3/i1D3,2D3
SMIL [2H]C([2H])([2H])c1cccc(c1N)C([2H])([2H])[2H]
IUPAC navn 2,6-bis(trideuteriomethyl)aniline
Molekylvægt (g/mol) 127.22
Formel vægt 127.1268
Procent renhed >95