Få mere at vide
2,6-Dimethylaniline-d11, TRC
Artikelnummer. 30373700
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30373700 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30373700 Leverandør Toronto Research Chemicals Leverandørnr. TRCD46219610MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2,6-Dimethylaniline-d11
CAS 1092805-08-7
Molekylær formel C8 D11 N
Mængde 10 mg
Synonym Benzen-3,4,5-d3-amine-d2, 2,6-di(methyl-d3)- (ACI), 2,6-Di(methyl-d3)benzen-3,4,5-d3-amine-d2 (ACI), 2,6-Dimethylaniline-d11, 2,6-Dimethylaniline-D11
InChI formel InChI=1S/C8H11N/c1-6-4-3-5-7(2)8(6)9/h3-5H,9H2,1-2H3/i1D3,2D3,3D,4D,5D/hD2
SMIL [2H]N([2H])c1c(c([2H])c([2H])c([2H])c1C([2H])([2H])[2H])C([2H])([2H])[2H]
IUPAC navn N,N,3,4,5-pentadeuterio-2,6-bis(trideuteriomethyl)aniline
Molekylvægt (g/mol) 132.2474
Formel vægt 132.1582