Få mere at vide
2,6-Dimethyl-d6-aniline, CDN
Artikelnummer. 30323987
Skift visning
Mængde:
0.25 g
0.5 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30323987 0.5 g 0.5g
30313022 0.25 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30323987 Leverandør CDN Leverandørnr. CDND65780.5G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2,6-Dimethylaniline-d6 (o-Xylidine-d6)
CAS 919785-81-2
Molekylær formel C8 2H6 H5 N
Mængde 0.5 g
InChI formel InChI=1 S/C8H11N/c1-6-4-3-5-7(2)8(6)9/h3-5 H,9H2,1-2H3/i1D3,2D3
SMIL [2 H]C([2 H])([2 H])c1cccc(c1N)C([2 H])([2 H])[2 H]
IUPAC navn 2,6-bis(trideuteriomethyl)aniline
Molekylvægt (g/mol) 127.22
Formel vægt 127.1268 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity