missing translation for 'onlineSavingsMsg'
Få mere at vide

2,6-Dimethoxytoluene, 98%

Artikelnummer. 11415197
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
25 g
100 g
Pakningsstørrelse:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
11415197 100 g 100g
11405197 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 11415197 Leverandør Thermo Scientific Alfa Aesar Leverandørnr. B22606.22
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter

Denne vare kan ikke returneres. Se returpolicy

CAS: 5673-07-4 | C9H12O2 | 152.19 g/mol

2,6-Dimethoxytoluene is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Dimethoxytoluene is used as an organic chemical synthesis intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 5673-07-4

Tekniske data

Kemisk navn eller materiale 2,6-Dimethoxytoluene
Smeltepunkt 37°C to 41°C
Kogepunkt 222°C
Flammepunkt 96°C (204°F)
Lugt Aromatic
Molekylær formel C9H12O2
MDL nummer MFCD00008374
Mængde 100 g
Beilstein 2045327
Synonym 2,6-dimethoxytoluene, benzene, 1,3-dimethoxy-2-methyl, 2-methyl-1,3-bis methyloxy benzene, 1,3-dimethoxy-2-methylbenzol, pubchem4545, acmc-1aqu0, 3-chloro-6-hydroxybiphenyl, 1,3-dimethoxy-2-methyl-benzene
Opløselighedsinformation Insoluble in water.
InChI nøgle FPEUDBGJAVKAEE-UHFFFAOYSA-N
SMIL CC1=C(C=CC=C1OC)OC
IUPAC navn 1,3-dimethoxy-2-methylbenzen
Molekylvægt (g/mol) 152.19
PubChem CID 79755
Formel vægt 152.19
Procent renhed 98%
Vis mere Vis mindre

RUO– Kun brug til forskning

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.