Få mere at vide
2,6-Diiodophenol, TRC
Artikelnummer. 30381204
Skift visning
Mængde:
10 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30381204 25 mg
30376973 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30381204 Leverandør Toronto Research Chemicals Leverandørnr. TRCD45892025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 28177-54-0
Molekylær formel C6 H4 I2 O
Molekylvægt (g/mol) 345.9
Synonym Phenol, 2,6-diiodo-, 2,6-Diiodophenol
IUPAC navn 2,6-diiodophenol
SMIL Oc1c(I)cccc1I
Tekniske data
Kemisk navn eller materiale 2,6-Diiodophenol
Mængde 25 mg
InChI formel InChI=1S/C6H4I2O/c7-4-2-1-3-5(8)6(4)9/h1-3,9H
Formel vægt 345.8352