missing translation for 'onlineSavingsMsg'
Learn More

2,6-Difluorobenzaldehyde, 97%, Thermo Scientific Chemicals

Catalog Number p-7023988
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
437-81-0
C7H4F2O
142.105
MFCD00010293
SOWRUJSGHKNOKN-UHFFFAOYSA-N
2,6-difluoro benzaldehyde, 2,6-dfad, benzaldehyde, 2,6-difluoro, 2,6-difluorobenzaledehyde, pubchem1438, 2,6-difluorbenzaldehyde, 2,6 difluorobenzaldehyde, acmc-1adi4, 2, 6-difluorobenzaldehyde, 2,6-difluoro-benzaldehyde
136284
2,6-difluorobenzaldehyde
C1=CC(=C(C(=C1)F)C=O)F
This item is not returnable. View return policy
437-81-0
C7H4F2O
142.105
MFCD00010293
SOWRUJSGHKNOKN-UHFFFAOYSA-N
2,6-difluoro benzaldehyde, 2,6-dfad, benzaldehyde, 2,6-difluoro, 2,6-difluorobenzaledehyde, pubchem1438, 2,6-difluorbenzaldehyde, 2,6 difluorobenzaldehyde, acmc-1adi4, 2, 6-difluorobenzaldehyde, 2,6-difluoro-benzaldehyde
136284
2,6-difluorobenzaldehyde
C1=CC(=C(C(=C1)F)C=O)F

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 15°C to 17°C
Density 1.317
Boiling Point 85°C to 86°C (20 mmHg)
Flash Point 73°C (163°F)
Refractive Index 1.507
Quantity 5 g
Beilstein 1935273
Sensitivity Air Sensitive
Formula Weight 142.11
Percent Purity 97%
Chemical Name or Material 2,6-Difluorobenzaldehyde
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'