missing translation for 'onlineSavingsMsg'
Learn More

2,6-Difluoro-N-(2-phenylethyl)benzamide, 97%, Thermo Scientific™

Catalog Number p-7039866
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
314054-21-2
C15H13F2NO
261.272
MFCD01175919
PORBEPWKQBWEGX-UHFFFAOYSA-N
2,6-difluoro-n-2-phenylethyl benzamide, 2,6-difluoro-n-phenethylbenzamide, 2,6-difluoro-n-phenethyl-benzamide, 2,6-difluoro-n-phenethylbenzenecarboxamide, 2,6-difluorophenyl-n-2-phenylethyl carboxamide
772213
2,6-difluoro-N-(2-phenylethyl)benzamide
C1=CC=C(C=C1)CCNC(=O)C2=C(C=CC=C2F)F
This item is not returnable. View return policy
314054-21-2
C15H13F2NO
261.272
MFCD01175919
PORBEPWKQBWEGX-UHFFFAOYSA-N
2,6-difluoro-n-2-phenylethyl benzamide, 2,6-difluoro-n-phenethylbenzamide, 2,6-difluoro-n-phenethyl-benzamide, 2,6-difluoro-n-phenethylbenzenecarboxamide, 2,6-difluorophenyl-n-2-phenylethyl carboxamide
772213
2,6-difluoro-N-(2-phenylethyl)benzamide
C1=CC=C(C=C1)CCNC(=O)C2=C(C=CC=C2F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 261.27
Percent Purity 97%
Chemical Name or Material 2,6-Difluoro-N-(2-phenylethyl)benzamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'