missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2,6-Dichlorothiophenol, 97%

Brand:  Alfa Aesar™ A12854.22

Additional Details : CAS Number : 24966-39-0 Weight : 0.10000kg

Quantity 100g

 View more versions of this product

Product Code. 11495263

  • 3731.00 DKK / 100g

Estimated Shipment
27-05-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

24966-39-0
179.058
JBISHCXLCGVPGW-UHFFFAOYSA-N
90671
C1=CC(=C(C(=C1)Cl)S)Cl
C6H4Cl2S
MFCD00004833
2,6-dichlorothiophenol, 2,6-dichlorobenzene-1-thiol, 2,6-dichloro thiophenol, benzenethiol, 2,6-dichloro, pubchem6802, 2,6-dichloro-benzenethiol, acmc-209gg5, benzenethiol,2,6-dichloro
2,6-dichlorobenzenethiol

Specifications

2,6-Dichlorothiophenol
Unpleasant
101°C to 103°C (14mmHg)
100g
C6H4Cl2S
2082400
JBISHCXLCGVPGW-UHFFFAOYSA-N
2,6-dichlorobenzenethiol
90671
97%
112°C (233°F)
Air Sensitive
46°C to 50°C
24966-39-0
MFCD00004833
2,6-dichlorothiophenol, 2,6-dichlorobenzene-1-thiol, 2,6-dichloro thiophenol, benzenethiol, 2,6-dichloro, pubchem6802, 2,6-dichloro-benzenethiol, acmc-209gg5, benzenethiol,2,6-dichloro
C1=CC(=C(C(=C1)Cl)S)Cl
179.058
179.07
Videos
SDS Documents