Få mere at vide
(2,6-Dichlorophenyl)acetic-alpha,alpha-d2 Acid, CDN
Artikelnummer. 30317245
Skift visning
Mængde:
0.25 g
0.5 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30317245 0.25 g -
30310012 0.5 g 0.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30317245 Leverandør CDN Leverandørnr. CDND71410.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2,6-Dichlorophenylacetic Acid-D2 (acetic-D2)
CAS 1219803-63-0
Molekylær formel C8 D2 H4 Cl2 O2
Mængde 0.25 g
Synonym Benzeneacetic-α,α-d2 acid, 2,6-dichloro- (ACI), 2,6-Dichlorobenzeneacetic-α,α-d2 acid (ACI), (2,6-Dichlorophenyl)acetic-α,α-d2 Acid, 2,6-Dichlorophenylacetic-d2 Acid, 2,6-Dichlorophenylacetic Acid-D2 (acetic-D2)
InChI formel InChI=1 S/C8H6Cl2O2/c9-6-2-1-3-7(10)5(6)4-8(11)12/h1-3 H,4H2,(H,11,12)/i4D2
SMIL [2 H]C([2 H])(C(=O)O)c1c(Cl)cccc1Cl
IUPAC navn 2,2-dideuterio-2-(2,6-dichlorophenyl)acetic acid
Molekylvægt (g/mol) 207.0504
Formel vægt 205.987 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity