missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichloroindophenol sodium salt, MP Biomedicals™

Catalog Number p-4404628
Click to view available options
Quantity:
1 g
This item is not returnable. View return policy
620-45-1
C12H6Cl2NNaO2
290.07
MFCD00150014
FHLDWQLHDYCXKI-UHFFFAOYSA-M
2,6-dichloroindophenol sodium salt, 2,6-dichlorophenolindophenol sodium salt, tillman's reagent, tillman's reagenz, dichlorphenol-indophenolnatrium, 2,6-dichloroindophenol sodium, unii-kad7q8xo1y, sodium 2,6-dichloroindophenol, sodium 2,6-dichloroindophenolate, 2,6-dichloroindophenol, sodium salt
23697355
CHEBI:948
sodium 2,6-dichloro-4-[(4-oxocyclohexa-2,5-dien-1-ylidene)amino]benzen-1-olate
[Na+].[O-]C1=C(Cl)C=C(C=C1Cl)N=C1C=CC(=O)C=C1
This item is not returnable. View return policy
620-45-1
C12H6Cl2NNaO2
290.07
MFCD00150014
FHLDWQLHDYCXKI-UHFFFAOYSA-M
2,6-dichloroindophenol sodium salt, 2,6-dichlorophenolindophenol sodium salt, tillman's reagent, tillman's reagenz, dichlorphenol-indophenolnatrium, 2,6-dichloroindophenol sodium, unii-kad7q8xo1y, sodium 2,6-dichloroindophenol, sodium 2,6-dichloroindophenolate, 2,6-dichloroindophenol, sodium salt
23697355
CHEBI:948
sodium 2,6-dichloro-4-[(4-oxocyclohexa-2,5-dien-1-ylidene)amino]benzen-1-olate
[Na+].[O-]C1=C(Cl)C=C(C=C1Cl)N=C1C=CC(=O)C=C1
Color Blue-Green
Quantity 1 g
Formula Weight 290.08
Physical Form Crystalline Powder
Chemical Name or Material 2, 6-Dichloroindophenol sodium salt
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'