missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichlorobenzene-1-sulfonyl chloride, ≥95%, Thermo Scientific™

Catalog Number p-2697264
Click to view available options
Quantity:
1 g
10 g
25 g
This item is not returnable. View return policy
6579-54-0
C6H3Cl3O2S
245.498
MFCD00052311
WGGKQIKICKLWGN-UHFFFAOYSA-N
2,6-dichlorobenzene-1-sulfonyl chloride, 2,6-dichlorobenzenesulfonylchloride, 2,6-dichlorophenylsulfonyl chloride, 2,6-dichlorobenzenesulphonyl chloride, unii-8523kf52zn, benzenesulfonyl chloride, 2,6-dichloro, 2,6-dichloro-benzenesulfonyl chloride, 2,6-dichlorobenzene sulfonyl chloride, pubchem5117, acmc-209ns8
522183
2,6-dichlorobenzenesulfonyl chloride
C1=CC(=C(C(=C1)Cl)S(=O)(=O)Cl)Cl
This item is not returnable. View return policy
6579-54-0
C6H3Cl3O2S
245.498
MFCD00052311
WGGKQIKICKLWGN-UHFFFAOYSA-N
2,6-dichlorobenzene-1-sulfonyl chloride, 2,6-dichlorobenzenesulfonylchloride, 2,6-dichlorophenylsulfonyl chloride, 2,6-dichlorobenzenesulphonyl chloride, unii-8523kf52zn, benzenesulfonyl chloride, 2,6-dichloro, 2,6-dichloro-benzenesulfonyl chloride, 2,6-dichlorobenzene sulfonyl chloride, pubchem5117, acmc-209ns8
522183
2,6-dichlorobenzenesulfonyl chloride
C1=CC(=C(C(=C1)Cl)S(=O)(=O)Cl)Cl
CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Linear Formula Cl2C6H3SO2Cl
Quantity 1 g
Formula Weight 245.51
Percent Purity 95%
Chemical Name or Material 2, 6-Dichlorobenzene-1-sulfonyl chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'