Få mere at vide
2,6-Dichlorobenzamide-3,4,5-d3, TRC
Artikelnummer. 30378867
Skift visning
Mængde:
2.5 mg
Pakningsstørrelse:
2.5mg
Artikelnummer. Mængde unitSize
30378867 2.5 mg 2.5mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30378867 Leverandør Toronto Research Chemicals Leverandørnr. TRCD4317462.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2,6-Dichlorobenzamide-3,4,5-d3
CAS 1219804-28-0
Molekylær formel C7 D3 H2 Cl2 N O
Mængde 2.5 mg
Synonym 2,6-Dichlorobenzamide D3 (2,6-Dichlorophenyl D3)
InChI formel InChI=1S/C7H5Cl2NO/c8-4-2-1-3-5(9)6(4)7(10)11/h1-3H,(H2,10,11)/i1D,2D,3D
SMIL [2H]c1c([2H])c(Cl)c(C(=O)N)c(Cl)c1[2H]
IUPAC navn 2,6-dichloro-3,4,5-trideuteriobenzamide
Molekylvægt (g/mol) 193.045
Formel vægt 191.994