Få mere at vide
2,6-Dichlorobenzamide-3,4,5-d3, CDN
Artikelnummer. 30326510
Skift visning
Mængde:
0.1 g
0.25 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30326510 0.1 g 0.1g
30312556 0.25 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30326510 Leverandør CDN Leverandørnr. CDND67000.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2,6-Dichlorobenzamide-3,4,5-d3
CAS 1219804-28-0
Molekylær formel C7 D3 H2 Cl2 N O
Mængde 0.1 g
Synonym 2,6-Dichlorobenzamide D3 (2,6-Dichlorophenyl D3)
InChI formel InChI=1 S/C7H5Cl2NO/c8-4-2-1-3-5(9)6(4)7(10)11/h1-3 H,(H2,10,11)/i1D,2 D,3 D
SMIL [2 H]c1c([2 H])c(Cl)c(C(=O)N)c(Cl)c1[2 H]
IUPAC navn 2,6-dichloro-3,4,5-trideuteriobenzamide
Molekylvægt (g/mol) 193.045
Formel vægt 191.994 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity