Learn More
2,6-Dichlorobenzaldehyde, TRC
Product Code. 30378831
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
Product Code. Quantity Unit Size
30378831 2.5 g 2.5g
1 options
Product Code. 30378831 Supplier Toronto Research Chemicals Supplier No. TRCD4318502.5G
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2,6-Dichlorobenzaldehyde
CAS 83-38-5
Molecular Formula C7H4Cl2O
Quantity 2.5 g
Synonym o,o'-Dichlorobenzaldehyde, NSC 7193
InChI Formula InChI=1S/C7H4Cl2O/c8-6-2-1-3-7(9)5(6)4-10/h1-4H
SMILES Clc1cccc(Cl)c1C=O
IUPAC Name 2,6-dichlorobenzaldehyde
Molecular Weight (g/mol) 175.01
Formula Weight 173.9639