Få mere at vide
2,6-Dichloro-alpha-(4-chlorophenyl)benzeneacetonitrile, TRC
Artikelnummer. 30137001
Skift visning
Mængde:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30137001 500 mg
30008302 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30137001 Leverandør Toronto Research Chemicals Leverandørnr. TRCD289180500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 85823-22-9
Molekylær formel C14 H8 Cl3 N
Molekylvægt (g/mol) 296.58
Synonym Benzeneacetonitrile, 2,6-dichloro-α-(4-chlorophenyl)-, 2,6-Dichloro-α-(4-chlorophenyl)benzeneacetonitrile, (RS)-(4-Chlorophenyl)(2,6-dichlorophenyl)acetonitrile, (RS)-2-(4-Chlorophenyl)-2-(2,6-dichlorophenyl)acetonitrile, Trichlorodiphenyl acetonitrile
IUPAC navn 2-(4-chlorophenyl)-2-(2,6-dichlorophenyl)acetonitrile
SMIL Clc1ccc(cc1)C(C#N)c2c(Cl)cccc2Cl
Tekniske data
Kemisk navn eller materiale 2,6-Dichloro-alpha-(4-chlorophenyl)benzeneacetonitrile
Analyt- eller komponentnavne 2,6-Dichloro-alpha-(4-chlorophenyl)benzeneacetonitrile
Forsendelsestilstand Room Temperature
InChI formel InChI=1S/C14H8Cl3N/c15-10-6-4-9(5-7-10)11(8-18)14-12(16)2-1-3-13(14)17/h1-7,11H
Løsningstype Neat
Industry Type Pharmaceutical
Mængde 500 mg
Formel vægt 294.9722