Learn More
2,6-Dichloro-4-(trifluoromethyl)phenylhydrazine, 97%, Thermo Scientific™
Product Code. 11490461
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11490461 5 g 5g
11480461 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11490461 Supplier Thermo Scientific Alfa Aesar Supplier No. B20013.06
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 86398-94-9
Molecular Formula C7H5Cl2F3N2
Molecular Weight (g/mol) 245.026
MDL Number MFCD00085001
InChI Key FYOWOHMZNWQLFG-UHFFFAOYSA-N
Synonym 2,6-dichloro-4-trifluoromethyl phenylhydrazine, 2,6-dichloro-4-trifluoromethyl phenyl hydrazine, 2,6-dichloro-4-trifluoro methylphenylhydrozine, 1-2,6-dichloro-4-trifluoromethyl phenyl hydrazine, pubchem4487, 3,5-dichloro-4-hydrazinobenzotrifluoride, 2,6-dichloro4-trifluoromethylphenylhydrazine, 2,6-dichloro-4-trifluoromethylphenylhydrazine, 2,6-dichloro-4-trifluoro methylphenylhydrazine
PubChem CID 688223
IUPAC Name [2,6-dichloro-4-(trifluoromethyl)phenyl]hydrazine
SMILES C1=C(C=C(C(=C1Cl)NN)Cl)C(F)(F)F
Specifications
Melting Point 57°C to 61°C
Quantity 5 g
Beilstein 8319619
Formula Weight 245.03
Percent Purity 97%
Chemical Name or Material 2,6-Dichloro-4-(trifluoromethyl)phenylhydrazine