Learn More
2,6-Dichloro-4-(trifluoromethoxy)aniline, 95%, BLD Pharm™
Specifications
Specifications
| CAS | 99479-66-0 |
| Molecular Formula | C7H4Cl2F3NO |
| MDL Number | MFCD00190128 |
| Quantity | 1 g |
| SMILES | NC1=C(Cl)C=C(OC(F)(F)F)C=C1Cl |
| Molecular Weight (g/mol) | 246.01 |
| Percent Purity | 95% |
| Chemical Name or Material | 2,6-Dichloro-4-(trifluoromethoxy)aniline |