Få mere at vide
2,6-Dibromophenol-3,4,5-d3, CDN
Artikelnummer. 30319850
Skift visning
Mængde:
0.05 g
0.1 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30319850 0.05 g -
30317858 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30319850 Leverandør CDN Leverandørnr. CDND43490.05G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2,6-Dibromophenol ring-d3
CAS 1219803-14-1
Molekylær formel C6 2H3 H Br2 O
Mængde 0.05 g
Synonym 2,6-Dibromophenol D3, 2,6-Dibromophenol (ring-D3), 2,6-Dibromophenol-3,4,5-D3, 2,6-Dibromo-3,4,5-trideuteriophenol D3
InChI formel InChI=1 S/C6H4Br2O/c7-4-2-1-3-5(8)6(4)9/h1-3,9 H/i1D,2 D,3 D
SMIL [2 H]c1c([2 H])c(Br)c(O)c(Br)c1[2 H]
IUPAC navn 2,6-dibromo-3,4,5-trideuteriophenol
Molekylvægt (g/mol) 254.92
Formel vægt 252.8817 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity