Learn More
2,6-Dibromo-4-isopropylaniline, 98%, BLD Pharm™
Product Code. 31031631
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31031631 5 g 5g
31016739 25 g 25g
31019861 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.
Product Code. 31031631 Supplier BLD Pharm Supplier No. BD112705g
 Restricted Item
Forventet forsendelse: 27-11-2026
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.
Specifications
CAS 10546-65-3
Molecular Formula C9H11Br2N
MDL Number MFCD00015475
Quantity 5 g
SMILES NC1=C(Br)C=C(C(C)C)C=C1Br
Molecular Weight (g/mol) 293.00
Percent Purity 98%
Chemical Name or Material 2,6-Dibromo-4-isopropylaniline