Få mere at vide
2,6-Diaminotoluene-d3, TRC
Artikelnummer. 30350752
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30350752 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30350752 Leverandør Toronto Research Chemicals Leverandørnr. TRCD41678725MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2,6-Diaminotoluene-d3
CAS 362049-58-9
Molekylær formel C7 2H3 H7 N2
Mængde 25 mg
Synonym 1,3-Benzenediamine, 2-(methyl-d3)- (9CI), 2-(Methyl-d3)-1,3-benzenediamine, 1,3-Diamino-2-(methyl-d3)benzene, 2,6-Diaminotoluene-d3, 2,6-Toluylenediamine-d3, 2-(Methyl-d3)-m-phenylenediamine, 2,6-Diaminotoluene-D3 (methyl-D3)
InChI formel InChI=1S/C7H10N2/c1-5-6(8)3-2-4-7(5)9/h2-4H,8-9H2,1H3/i1D3
SMIL [2H]C([2H])([2H])c1c(N)cccc1N
IUPAC navn 2-(trideuteriomethyl)benzene-1,3-diamine
Molekylvægt (g/mol) 125.19
Formel vægt 125.1032