missing translation for 'onlineSavingsMsg'
Learn More

2,6-Diaminotoluene, 98%, Thermo Scientific Chemicals

Catalog Number p-100375
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
823-40-5
C7H10N2
122.17
MFCD00007800
RLYCRLGLCUXUPO-UHFFFAOYSA-N
2,6-diaminotoluene, 2,6-toluenediamine, 1,3-benzenediamine, 2-methyl, toluene-2,6-diamine, 2,6-tolylenediamine, 2,6-toluylenediamine, 2,6-diamino-1-methylbenzene, 2-methyl-1,3-benzenediamine, 2-methyl-1,3-phenylenediamine, 2-methyl-m-phenylenediamine
13205
CHEBI:76288
2-methylbenzene-1,3-diamine
CC1=C(N)C=CC=C1N
This item is not returnable. View return policy
823-40-5
C7H10N2
122.17
MFCD00007800
RLYCRLGLCUXUPO-UHFFFAOYSA-N
2,6-diaminotoluene, 2,6-toluenediamine, 1,3-benzenediamine, 2-methyl, toluene-2,6-diamine, 2,6-tolylenediamine, 2,6-toluylenediamine, 2,6-diamino-1-methylbenzene, 2-methyl-1,3-benzenediamine, 2-methyl-1,3-phenylenediamine, 2-methyl-m-phenylenediamine
13205
CHEBI:76288
2-methylbenzene-1,3-diamine
CC1=C(N)C=CC=C1N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 102.0°C to 107.0°C
Color Black or Brown to Gray
Boiling Point 289.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula CH3C6H3(NH2)2
Quantity 5 g
Beilstein 13,148
Solubility Information Solubility in water: 60g/L water (15°C).
Formula Weight 122.17
Percent Purity 98%
Physical Form Pellets, Powder or Chunks
Chemical Name or Material 2, 6-Diaminotoluene, 98%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2,6-Diaminotoluene
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Germ cell mutagenicity Category 2
  • Respiratory sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H341-Suspected of causing genetic defects.
  • H411-Toxic to aquatic life with long lasting effects.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'