Få mere at vide
2,6-Di-(tert-butyl-d9)-4-methylphenol-3,5-d2,OD, TRC
Artikelnummer. 30376822
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30376822 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30376822 Leverandør Toronto Research Chemicals Leverandørnr. TRCD4294865MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Butylhydroxytoluene-d21
CAS 64502-99-4
Renhedsgrad noter HPLC
Molekylær formel C15 2H21 H3 O
Mængde 5 mg
Synonym Phen-3,5-d2-ol-d, 2,6-bis[1,1-di(methyl-d3)ethyl-2,2,2-d3]-4-methyl- (9CI), Butylhydroxytoluene-d21, d21-2,6-di(tert-butyl)-4-methylphenol, BHT-d21, 2,6-Di-tert-butyl-4-methylphenol D21
InChI formel InChI=1S/C15H24O/c1-10-8-11(14(2,3)4)13(16)12(9-10)15(5,6)7/h8-9,16H,1-7H3/i2D3,3D3,4D3,5D3,6D3,7D3,8D,9D/hD
SMIL [2H]Oc1c(c([2H])c(C)c([2H])c1C(C([2H])([2H])[2H])(C([2H])([2H])[2H])C([2H])([2H])[2H])C(C([2H])([2H])[2H])(C([2H])([2H])[2H])C([2H])([2H])[2H]
IUPAC navn 1,5-dideuterio-3-deuteriooxy-2,4-bis[1,1,1,3,3,3-hexadeuterio-2-(trideuteriomethyl)propan-2-yl]-6-methylbenzene
Molekylvægt (g/mol) 241.48
Formel vægt 241.3145
Procent renhed >95