Få mere at vide
2,6-Di-(tert-butyl-d9)-4-methylphenol-3,5-d2,OD, CDN
Artikelnummer. 30311022
Skift visning
Mængde:
0.1 g
0.25 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30311022 0.1 g 0.1g
30309831 0.25 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30311022 Leverandør CDN Leverandørnr. CDND17650.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Butylhydroxytoluene-d21
CAS 64502-99-4
Molekylær formel C15 2H21 H3 O
Mængde 0.1 g
Synonym Phen-3,5-d2-ol-d, 2,6-bis[1,1-di(methyl-d3)ethyl-2,2,2-d3]-4-methyl- (9 CI), Butylhydroxytoluene-d21, d21-2,6-di(tert-butyl)-4-methylphenol, BHT-d21
InChI formel InChI=1 S/C15H24O/c1-10-8-11(14(2,3)4)13(16)12(9-10)15(5,6)7/h8-9,16 H,1-7H3/i2D3,3D3,4D3,5D3,6D3,7D3,8 D,9 D/hD
SMIL [2 H]Oc1c(c([2 H])c(C)c([2 H])c1C(C([2 H])([2 H])[2 H])(C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H])C(C([2 H])([2 H])[2 H])(C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H]
IUPAC navn 1,5-dideuterio-3-deuteriooxy-2,4-bis[1,1,1,3,3,3-hexadeuterio-2-(trideuteriomethyl)propan-2-yl]-6-methylbenzene
Molekylvægt (g/mol) 241.48
Formel vægt 241.3145 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity