Få mere at vide
2,6-Di-tert-butyl-4-methylphenol-d24, TRC
Artikelnummer. 30378855
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30378855 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30378855 Leverandør Toronto Research Chemicals Leverandørnr. TRCD42948710MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2,6-Di-tert-butyl-4-methylphenol-d24
CAS 1219805-92-1
Molekylær formel C15 2H24 O
Mængde 10 mg
Synonym 2,6-Di-tert-butyl-4-methylphenol D24, Phen-3,5-d2-ol-d, 2,6-bis[1,1-di(methyl-d3)ethyl-2,2,2-d3]-4-(methyl-d3)-, 2,6-Di-tert-butyl-4-methylphenol-d24
InChI formel InChI=1S/C15H24O/c1-10-8-11(14(2,3)4)13(16)12(9-10)15(5,6)7/h8-9,16H,1-7H3/i1D3,2D3,3D3,4D3,5D3,6D3,7D3,8D,9D/hD
SMIL [2H]Oc1c(c([2H])c(c([2H])c1C(C([2H])([2H])[2H])(C([2H])([2H])[2H])C([2H])([2H])[2H])C([2H])([2H])[2H])C(C([2H])([2H])[2H])(C([2H])([2H])[2H])C([2H])([2H])[2H]
IUPAC navn 1,5-dideuterio-3-deuteriooxy-2,4-bis[1,1,1,3,3,3-hexadeuterio-2-(trideuteriomethyl)propan-2-yl]-6-(trideuteriomethyl)benzene
Molekylvægt (g/mol) 244.5
Formel vægt 244.3334