Få mere at vide
2,6-Di-tert-butyl-4-methylphenol D21, Dr. Ehrenstorfer
Artikelnummer. 30310011
Skift visning
Mængde:
25 mg
Pakningsstørrelse:
25mg
Artikelnummer. Mængde unitSize
30310011 25 mg 25mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30310011 Leverandør Dr. Ehrenstorfer Leverandørnr. DREC12253501
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Butylhydroxytoluene-d21
CAS 64502-99-4
Molekylær formel C15 2H21 H3 O
Mængde 25 mg
Synonym Phen-3,5-d2-ol-d, 2,6-bis[1,1-di(methyl-d3)ethyl-2,2,2-d3]-4-methyl- (9CI), Butylhydroxytoluene-d21, d21-2,6-di(tert-butyl)-4-methylphenol, BHT-d21
InChI formel InChI=1S/C15H24O/c1-10-8-11(14(2,3)4)13(16)12(9-10)15(5,6)7/h8-9,16H,1-7H3/i2D3,3D3,4D3,5D3,6D3,7D3,8D,9D/hD
SMIL [2H]Oc1c(c([2H])c(C)c([2H])c1C(C([2H])([2H])[2H])(C([2H])([2H])[2H])C([2H])([2H])[2H])C(C([2H])([2H])[2H])(C([2H])([2H])[2H])C([2H])([2H])[2H]
IUPAC navn 1,5-dideuterio-3-deuteriooxy-2,4-bis[1,1,1,3,3,3-hexadeuterio-2-(trideuteriomethyl)propan-2-yl]-6-methylbenzene
Molekylvægt (g/mol) 241.48
Formel vægt 241.3145 g/mol