Få mere at vide
2,6,10,14-Tetramethylpentadecane-d40, CDN
Artikelnummer. 30320607
Skift visning
Mængde:
0.25 g
0.5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30320607 0.5 g
30313940 0.25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30320607 Leverandør CDN Leverandørnr. CDND09090.5G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Pristane-D40
CAS 16416-35-6
Molekylær formel C19 D40
Mængde 0.5 g
Synonym Pentadecane-1,1,1,2,3,3,4,4,5,5,6,7,7,8,8,9,9,10,11,11,12,12,13,13,14,15,15,15-d28, 2,6,10,14-tetra(methyl-d3)- (8 CI, 9 CI), 2,6,10,14-Tetramethylpentadecane-d40, Pristane-D40, Norphytane-d40
InChI formel InChI=1 S/C19H40/c1-16(2)10-7-12-18(5)14-9-15-19(6)13-8-11-17(3)4/h16-19 H,7-15H2,1-6H3/i1D3,2D3,3D3,4D3,5D3,6D3,7D2,8D2,9D2,10D2,11D2,12D2,13D2,14D2,15D2,16 D,17 D,18 D,19 D
SMIL [2 H]C([2 H])([2 H])C([2 H])(C([2 H])([2 H])[2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])(C([2 H])([2 H])[2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])(C([2 H])([2 H])[2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])(C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H]
IUPAC navn 1,1,1,2,3,3,4,4,5,5,6,7,7,8,8,9,9,10,11,11,12,12,13,13,14,15,15,15-octacosadeuterio-2,6,10,14-tetrakis(trideuteriomethyl)pentadecane
Molekylvægt (g/mol) 308.7674
Formel vægt 308.5641 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity