Få mere at vide
2-[[(5-Methyl-1H-imidazol-4-yl)methyl]sulphanyl]ethanamine Dihydrobromide, Mikromol™
Beskrivelse
2-[[(5-Methyl-1H-imidazol-4-yl)methyl]sulphanyl]ethanamine Dihydrobromide
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 2-[((5-Methyl-1H-imidazol-4-yl)methyl)sulphanyl]ethanamine Dihydrobromide |
| CAS | 38603-73-5 |
| Analyt- eller komponentnavne | 2-[((5-Methyl-1H-imidazol-4-yl)methyl)sulphanyl]ethanamine Dihydrobromide |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C7 H13 N3 S . 2 Br H |
| Certificeringer/compliance | ISO 17025 |
| Synonym | 2-[[(5-Methyl-1H-imidazol-4-yl)methyl]sulphanyl]ethanamine Dihydrobromide, Cimetidine Hydrochloride Imp. J (EP) as Dihydrobromide, Cimetidine Hydrochloride Imp. J (EP), Cimetidine Imp. J (EP), Cimetidine Hydrochloride Impurity J, Cimetidine Impurity J as Dihydrobromide, Cimetidine Impurity J |
| InChI formel | InChI=1S/C7H13N3S.2BrH/c1-6-7(10-5-9-6)4-11-3-2-8;;/h5H,2-4,8H2,1H3,(H,9,10);2*1H |
| SMIL | Br.Br.Cc1[nH]cnc1CSCCN |
| IUPAC navn | 2-[(5-methyl-1H-imidazol-4-yl)methylsulfanyl]ethanamine;dihydrobromide |
| Vis mere |