Få mere at vide
2-(5-Methoxybenzo[d]isoxazol-3-yl)acetic Acid, TRC
Artikelnummer. 30480644
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30480644 50 mg
30472009 100 mg
30472202 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30480644 Leverandør Toronto Research Chemicals Leverandørnr. TRCM26671550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 34172-98-0
Molekylær formel C10H9NO4
Molekylvægt (g/mol) 207.18
Synonym 2-(5-Methoxy-1,2-benzoxazol-3-yl)acetic acid
IUPAC navn 2-(5-methoxy-1,2-benzoxazol-3-yl)acetic acid
SMIL COc1ccc2onc(CC(=O)O)c2c1
Tekniske data
Kemisk navn eller materiale 2-(5-Methoxybenzo[d]isoxazol-3-yl)acetic Acid
Mængde 50 mg
InChI formel InChI=1S/C10H9NO4/c1-14-6-2-3-9-7(4-6)8(11-15-9)5-10(12)13/h2-4H,5H2,1H3,(H,12,13)
Formel vægt 207.0532