Få mere at vide
2-(5-Methoxy-1H-indol-3-yl)ethanamine (5-Methoxytryptamine), Mikromol™
Artikelnummer. 30050351
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30050351 25 mg
17963656 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30050351 Leverandør Mikromol Leverandørnr. MM1373.010025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2-(5-Methoxy-1H-indol-3-yl)ethanamine (5-Methoxytryptamine)
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 5-Methoxytryptamine
CAS 608-07-1
Analyt- eller komponentnavne 5-Methoxytryptamine
Forsendelsestilstand Room Temperature
Molekylær formel C11 H14 N2 O
Certificeringer/compliance ISO 17025
Synonym 2-(5-Methoxy-1H-indol-3-yl)ethanamine, 5-Methoxytryptamine, Melatonin Related Compound A (USP), Melatonin Related Compound A
InChI formel InChI=1S/C11H14N2O/c1-14-9-2-3-11-10(6-9)8(4-5-12)7-13-11/h2-3,6-7,13H,4-5,12H2,1H3
SMIL COc1ccc2[nH]cc(CCN)c2c1
IUPAC navn 2-(5-methoxy-1H-indol-3-yl)ethanamine
Molekylvægt (g/mol) 190.24
Mængde 25 mg
Formel vægt 190.1106 g/mol