Få mere at vide
2-(5-Mercaptotetrazole-1-yl)ethanol, TRC
Artikelnummer. 30475113
Skift visning
Mængde:
1 g
5 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30475113 1 g
30474042 50 g
30472262 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30475113 Leverandør Toronto Research Chemicals Leverandørnr. TRCM2254601G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 56610-81-2
Molekylær formel C3 H6 N4 O S
Molekylvægt (g/mol) 146.17
Synonym 1,2-Dihydro-1-(2-hydroxyethyl)-5H-tetrazole-5-thione, 1-(2-Hydroxyethyl)-1H-tetrazole-5-thiol, 1-(2-Hydroxyethyl)-5-mercapto-1H-tetrazole, 1-(2-Hydroxyethyl)-5-mercaptotetrazole, 1-(2-Hydroxyethyl)tetrazole-5-thiol, 5-Mercapto-1-(2-hydroxyethyl)-1H-tetrazole, 5-Mercapto-1-tetrazoleethanol, [1-(2-Hydroxyethyl)-1H-1H-tetrazol-5-yl]thiol
IUPAC navn 2-(5-sulfanyltetrazol-1-yl)ethanol
SMIL OCCn1nnnc1S
Tekniske data
Kemisk navn eller materiale 2-(5-Mercaptotetrazole-1-yl)ethanol
Mængde 1 g
InChI formel InChI=1S/C3H6N4OS/c8-2-1-7-3(9)4-5-6-7/h8H,1-2H2,(H,4,6,9)
Formel vægt 146.0262