Få mere at vide
2,5-Hexanedione-d10, CDN
Artikelnummer. 30328474
Skift visning
Mængde:
0.1 g
0.25 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30328474 0.25 g -
30309028 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30328474 Leverandør CDN Leverandørnr. CDND22960.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2,5-Hexanedione-d10
CAS 97135-07-4
Molekylær formel C6 D10 O2
Mængde 0.25 g
Synonym 2,5-Hexanedione-1,1,1,3,3,4,4,6,6,6-d10 (9 CI), 2,5-Hexanedione-d10, 2,5-Hexanedione D10, 2,5-Diketohexane-d10, Acetonylacetone-d10, Diacetonyl-d10
InChI formel InChI=1 S/C6H10O2/c1-5(7)3-4-6(2)8/h3-4H2,1-2H3/i1D3,2D3,3D2,4D2
SMIL [2 H]C([2 H])([2 H])C(=O)C([2 H])([2 H])C([2 H])([2 H])C(=O)C([2 H])([2 H])[2 H]
IUPAC navn 1,1,1,3,3,4,4,6,6,6-decadeuteriohexane-2,5-dione
Molekylvægt (g/mol) 124.204
Formel vægt 124.131 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity