Få mere at vide
2,5-Dinitrotoluene, TRC
Artikelnummer. 30133640
Skift visning
Mængde:
100 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30133640 1 g
30137194 250 mg
30008771 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30133640 Leverandør Toronto Research Chemicals Leverandørnr. TRCD4268101G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 619-15-8
Molekylær formel C7 H6 N2 O4
Molekylvægt (g/mol) 182.13
Synonym Toluene, 2,5-dinitro- (8CI), 2-Methyl-1,4-dinitrobenzene, 1-Methyl-2,5-dinitrobenzene, 2,5-DNT, 2,5-Dinitrotoluene, 3-Methyl-1,4-dinitrobenzene, NSC 159120, Benzene, 2-methyl-1,4-dinitro-
IUPAC navn 2-methyl-1,4-dinitrobenzene
SMIL Cc1cc(ccc1[N+](=O)[O-])[N+](=O)[O-]
Tekniske data
Kemisk navn eller materiale 2,5-Dinitrotoluene
Renhedsgrad noter HPLC
Mængde 1 g
InChI formel InChI=1S/C7H6N2O4/c1-5-4-6(8(10)11)2-3-7(5)9(12)13/h2-4H,1H3
Formel vægt 182.0328
Procent renhed >95%