missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethylthiophene, 98+%, Thermo Scientific Chemicals

Catalog Number p-7081019
Click to view available options
Quantity:
10 g
25 g
This item is not returnable. View return policy
638-02-8
C6H8S
112.19
MFCD00005452
GWQOOADXMVQEFT-UHFFFAOYSA-N
thiophene, 2,5-dimethyl, 2,5-dimethyl-thiophene, unii-v6ddx6wb12, v6ddx6wb12, pubchem5194, 2,5 dimethylthiophene, thiophene,5-dimethyl, 2,5-dimethyl thiophene, thiophene,2,5-dimethyl, 2,5-dimethyl-2h-thiolium
12514
2,5-dimethylthiophene
CC1=CC=C(S1)C
This item is not returnable. View return policy
638-02-8
C6H8S
112.19
MFCD00005452
GWQOOADXMVQEFT-UHFFFAOYSA-N
thiophene, 2,5-dimethyl, 2,5-dimethyl-thiophene, unii-v6ddx6wb12, v6ddx6wb12, pubchem5194, 2,5 dimethylthiophene, thiophene,5-dimethyl, 2,5-dimethyl thiophene, thiophene,2,5-dimethyl, 2,5-dimethyl-2h-thiolium
12514
2,5-dimethylthiophene
CC1=CC=C(S1)C

2,5-Dimethylthiophene is used as an intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,5-Dimethylthiophene is used as an intermediate in organic synthesis.

Solubility
Insoluble in water. Soluble in alcohol, ether and benzene.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point -63°C
Density 0.984
Boiling Point 134°C to 136°C
Flash Point 24°C (75°F)
Odor Characteristic
Refractive Index 1.512
Quantity 10 g
UN Number UN1993
Beilstein 106450
Solubility Information Insoluble in water. Soluble in alcohol,ether and benzene.
Formula Weight 112.19
Percent Purity ≥98%
Chemical Name or Material 2,5-Dimethylthiophene
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'