Learn More
2,5-Dimethylfuran-3-carbonyl chloride, 97%, Maybridge™
Brand: Maybridge CC01102DE
Additional Details : CAS Number : 50990-93-7 Weight : 0.05500kg Transport : UN number : 3265 Chem class : 8 Pack group : II
Quantity | 5g |
---|
Chemical Identifiers
50990-93-7 | |
158.581 | |
RWORXZHVOUPMMM-UHFFFAOYSA-N | |
2734804 | |
CC1=CC(=C(O1)C)C(=O)Cl |
C7H7ClO2 | |
MFCD00221073 | |
2,5-dimethyl-3-furoyl chloride, 2,5-dimethyl-furan-3-carbonyl chloride, 2,5-dimethyl-3-furancarbonyl chloride, 2,5-dimethylfuran-3-carbonylchloride, pubchem6936, acmc-209krc, 2,5-dimethyl-3-furoylchloride, 2,5 dimethyl-furan-3-carbonyl chloride, 2,5-dimethyl furan-3-carbonyl chloride, 3-furancarbonylchloride, 2,5-dimethyl | |
2,5-dimethylfuran-3-carbonyl chloride |
Specifications
2, 5-Dimethylfuran-3-carbonyl chloride | |
5g | |
96.5 | |
97% | |
MFCD00221073 | |
RWORXZHVOUPMMM-UHFFFAOYSA-N | |
2,5-dimethylfuran-3-carbonyl chloride | |
2734804 | |
97% |
1.19g/mL | |
50990-93-7 | |
100.0 | |
C7H7ClO2 | |
2,5-dimethyl-3-furoyl chloride, 2,5-dimethyl-furan-3-carbonyl chloride, 2,5-dimethyl-3-furancarbonyl chloride, 2,5-dimethylfuran-3-carbonylchloride, pubchem6936, acmc-209krc, 2,5-dimethyl-3-furoylchloride, 2,5 dimethyl-furan-3-carbonyl chloride, 2,5-dimethyl furan-3-carbonyl chloride, 3-furancarbonylchloride, 2,5-dimethyl | |
CC1=CC(=C(O1)C)C(=O)Cl | |
158.581 | |
158.58 |