Få mere at vide
2,5-Dimethyl Celecoxib, TRC
Artikelnummer. 30383219
Skift visning
Mængde:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30383219 250 mg
30374066 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30383219 Leverandør Toronto Research Chemicals Leverandørnr. TRCD470890250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 457639-26-8
Molekylær formel C18 H16 F3 N3 O2 S
Molekylvægt (g/mol) 395.3987
Synonym 2,5-Dimethylcelecoxib, Benzenesulfonamide, 4-[5-(2,5-dimethylphenyl)-3-(trifluoromethyl)-1H-pyrazol-1-yl]-, 4-[5-(2,5-Dimethylphenyl)-3-(trifluoromethyl)-1H-pyrazol-1-yl]benzenesulfonamide
IUPAC navn 4-[5-(2,5-dimethylphenyl)-3-(trifluoromethyl)pyrazol-1-yl]benzenesulfonamide
SMIL Cc1ccc(C)c(c1)c2cc(nn2c3ccc(cc3)S(=O)(=O)N)C(F)(F)F
Tekniske data
Kemisk navn eller materiale 2,5-Dimethylcelecoxib
Mængde 250 mg
InChI formel InChI=1S/C18H16F3N3O2S/c1-11-3-4-12(2)15(9-11)16-10-17(18(19,20)21)23-24(16)13-5-7-14(8-6-13)27(22,25)26/h3-10H,1-2H3,(H2,22,25,26)
Formel vægt 395.0915