Få mere at vide
2,5-Diisopropylphenol, TRC
Artikelnummer. 30008066
Skift visning
Mængde:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30008066 250 mg
30005893 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30008066 Leverandør Toronto Research Chemicals Leverandørnr. TRCD455390250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 35946-91-9
Molekylær formel C12 H18 O
Molekylvægt (g/mol) 178.27
Synonym 2,5-Bis(1-methylethyl)phenol, 2,5-Diisopropylphenol, Propofol Imp. D (EP)
IUPAC navn 2,5-di(propan-2-yl)phenol
SMIL CC(C)c1ccc(C(C)C)c(O)c1
Tekniske data
Kemisk navn eller materiale 2,5-Diisopropylphenol
Mængde 250 mg
InChI formel InChI=1S/C12H18O/c1-8(2)10-5-6-11(9(3)4)12(13)7-10/h5-9,13H,1-4H3
Formel vægt 178.1358