Få mere at vide
(2,5-Difluorophenyl)-(4-piperidyl)methanone Hydrochloride, Mikromol
Artikelnummer. 30462414
Skift visning
Mængde:
25 mg
4 x 25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30462414 4 x 25 mg
30049598 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30462414 Leverandør Mikromol Leverandørnr. MM3659.0100254AMP
Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
(2,5-Difluorophenyl)-(4-piperidyl)methanone Hydrochloride, Mikromol
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (2,5-Difluorophenyl)-(4-piperidyl)methanone Hydrochloride
CAS 1172297-96-9
Analyt- eller komponentnavne (2,5-Difluorophenyl)-(4-piperidyl)methanone Hydrochloride
Forsendelsestilstand Room Temperature
Molekylær formel C12 H13 F2 N O . H Cl
Certificeringer/compliance ISO 17025
Synonym (2,5-Difluorophenyl)-(4-piperidyl)methanone Hydrochloride
InChI formel InChI=1S/C12H13F2NO.ClH/c13-9-1-2-11(14)10(7-9)12(16)8-3-5-15-6-4-8;/h1-2,7-8,15H,3-6H2;1H
Løsningstype Neat
SMIL Cl.Fc1ccc(F)c(c1)C(=O)C2CCNCC2
IUPAC navn (2,5-difluorophenyl)-piperidin-4-ylmethanone;hydrochloride
Molekylvægt (g/mol) 261.7
Mængde 4 x 25 mg
Formel vægt 261.696 g/mol