missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2,5-Dichloronicotinic acid, 98%

Brand:  Alfa Aesar™ H64998.06

Additional Details : CAS Number : 59782-85-3 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15472439

  • 1006.82 DKK / 5g


In stock, ready to ship: 1
Add to basket

Specifications

Chemical Identifiers

59782-85-3
191.995
SXQSMLIMBNMUNB-UHFFFAOYSA-N
2736085
C1=C(C(=NC=C1Cl)Cl)C(=O)O
C6H3Cl2NO2
MFCD01861979
2,5-dichloronicotinic acid, 2,5-dichloronicotinicacid, 3-pyridinecarboxylic acid, 2,5-dichloro, 2,5-dichloro-3-pyridinecarboxylic acid, pubchem2601, acmc-209mfn, pyni16, 2,5-dichloro-nicotinic acid, 2,5-dichloronicotinic acid?, ksc269g2r
2,5-dichloropyridine-3-carboxylic acid

Specifications

2,5-Dichloronicotinic acid
White
5g
C6H3Cl2NO2
389586
SXQSMLIMBNMUNB-UHFFFAOYSA-N
2,5-dichloropyridine-3-carboxylic acid
2736085
Powder
1.61 g/mL
156°C to 158°C
59782-85-3
MFCD01861979
2,5-dichloronicotinic acid, 2,5-dichloronicotinicacid, 3-pyridinecarboxylic acid, 2,5-dichloro, 2,5-dichloro-3-pyridinecarboxylic acid, pubchem2601, acmc-209mfn, pyni16, 2,5-dichloro-nicotinic acid, 2,5-dichloronicotinic acid?, ksc269g2r
C1=C(C(=NC=C1Cl)Cl)C(=O)O
191.995
192
98%
Videos
SDS Documents