missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dibromothiophene, 95%, Thermo Scientific Chemicals

Catalog Number p-7021177
Click to view available options
Quantity:
25 g
50 g
250 g
This item is not returnable. View return policy
3141-27-3
C4H2Br2S
241.93
MFCD00005420
KBVDUUXRXJTAJC-UHFFFAOYSA-N
thiophene, 2,5-dibromo, 2,5-dibromo-thiophene, unii-f0033k8cxm, pubchem5512, thiophene,5-dibromo, acmc-1ckmh, 2,5-bis bromanyl thiophene, ksc224g0j, 2,5-dibromothiophene, bidd:gt0086
18453
2,5-dibromothiophene
BrC1=CC=C(Br)S1
This item is not returnable. View return policy
3141-27-3
C4H2Br2S
241.93
MFCD00005420
KBVDUUXRXJTAJC-UHFFFAOYSA-N
thiophene, 2,5-dibromo, 2,5-dibromo-thiophene, unii-f0033k8cxm, pubchem5512, thiophene,5-dibromo, acmc-1ckmh, 2,5-bis bromanyl thiophene, ksc224g0j, 2,5-dibromothiophene, bidd:gt0086
18453
2,5-dibromothiophene
BrC1=CC=C(Br)S1

2,5-Dibromothiophene, is mainly used as pharmaceutical intermediate and synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,5-Dibromothiophene, is mainly used as pharmaceutical intermediate and synthesis intermediate.

Solubility
It is soluble in chloroform and methanol. It is insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Strong oxidizing agents.Light Sensitive
TRUSTED_SUSTAINABILITY
Melting Point -6°C
Density 2.15
Boiling Point 210°C to 212°C
Flash Point 99°C (210°F)
Odor Pungent
Refractive Index 1.629
Quantity 25 g
UN Number UN2810
Beilstein 109899
Solubility Information It is soluble in chloroform and methanol. It is insoluble in water.
Formula Weight 241.94
Percent Purity 96%
Chemical Name or Material 2,5-Dibromothiophene
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'