missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dibromonitrobenzene, 97%, Thermo Scientific Chemicals

Catalog Number p-102781
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
3460-18-2
C6H3Br2NO2
280.89
MFCD00007046
WRGKKASJBOREMB-UHFFFAOYSA-N
2,5-dibromonitrobenzene, 1,4-dibromo-2-nitro-benzene, benzene, 1,4-dibromo-2-nitro, 2,5-dibromo-1-nitrobenzene, rarechem fh 1w 0042, pubchem3899, 1,4-dibromonitrobenzene, 2,5-dibromo nitrobenzene, 2,5-dibromo-nitrobenzene, acmc-1ahj1
77004
1,4-dibromo-2-nitrobenzene
C1=CC(=C(C=C1Br)[N+](=O)[O-])Br
This item is not returnable. View return policy
3460-18-2
C6H3Br2NO2
280.89
MFCD00007046
WRGKKASJBOREMB-UHFFFAOYSA-N
2,5-dibromonitrobenzene, 1,4-dibromo-2-nitro-benzene, benzene, 1,4-dibromo-2-nitro, 2,5-dibromo-1-nitrobenzene, rarechem fh 1w 0042, pubchem3899, 1,4-dibromonitrobenzene, 2,5-dibromo nitrobenzene, 2,5-dibromo-nitrobenzene, acmc-1ahj1
77004
1,4-dibromo-2-nitrobenzene
C1=CC(=C(C=C1Br)[N+](=O)[O-])Br

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 81°C to 85°C
Color Yellow
Density 2.3740g/mL
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula Br2C6H3NO2
Quantity 25 g
Beilstein 05, 250
Specific Gravity 2.374
Formula Weight 280.89
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 2, 5-Dibromonitrobenzene, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 2,5-Dibromonitrobenzene
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'