Få mere at vide
2,5-Di-(tert-butyl-d9)-4-methoxyphenol-3,6-d2, CDN
Artikelnummer. 30318297
Skift visning
Mængde:
0.01 g
0.05 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30318297 0.01 g
30319014 0.05 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30318297 Leverandør CDN Leverandørnr. CDND62740.01G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2,5-Di-tert-butyl-4-methoxyphenol-D20 (ditertbutyl-D18,phenol-3,6-D2)
CAS 1219802-07-9
Molekylær formel C15 D20 H4 O2
Mængde 0.01 g
Synonym 2,5-Di[1,1-di(methyl-d3)ethyl-2,2,2-d3]-4-methoxyphen-3,6-d2-ol, 2,5-Di-tert-butyl-4-hydroxyanisole-d20, 2,5-Di-tert-butyl-4-methoxyphenol-D20 (ditertbutyl-D18,phenol-3,6-D2), DTBMP-d20, 2,5-Di-(tert-butyl-d9)-4-methoxyphenol-3,6-d2
InChI formel InChI=1 S/C15H24O2/c1-14(2,3)10-9-13(17-7)11(8-12(10)16)15(4,5)6/h8-9,16 H,1-7H3/i1D3,2D3,3D3,4D3,5D3,6D3,8 D,9 D
SMIL [2 H]c1c(O)c(c([2 H])c(OC)c1C(C([2 H])([2 H])[2 H])(C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H])C(C([2 H])([2 H])[2 H])(C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H]
IUPAC navn 2,5-dideuterio-3,6-bis[1,1,1,3,3,3-hexadeuterio-2-(trideuteriomethyl)propan-2-yl]-4-methoxyphenol
Molekylvægt (g/mol) 256.4731
Formel vægt 256.3032 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity