Få mere at vide
2-(((5-Chloro-2-(methylamino)phenyl)(phenyl)methylene)amino)ethanol, TRC
Artikelnummer. 30368600
Skift visning
Mængde:
25 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30368600 25 mg
30351973 250 mg
30352244 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30368600 Leverandør Toronto Research Chemicals Leverandørnr. TRCC38735025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 2271-06-9
Molekylær formel C16H17ClN2O
Molekylvægt (g/mol) 288.77
Synonym MAB, 2-Methylamino-5-chlor-alpha-phenylbenzylidenaminoethanol
IUPAC navn 2-[(Z)-[[5-chloro-2-(methylamino)phenyl]-phenylmethylidene]amino]ethanol
SMIL CNc1ccc(Cl)cc1\C(=N/CCO)\c2ccccc2
Tekniske data
Kemisk navn eller materiale 2-(((5-Chloro-2-(methylamino)phenyl)(phenyl)methylene)amino)ethanol
Mængde 25 mg
InChI formel InChI=1S/C16H17ClN2O/c1-18-15-8-7-13(17)11-14(15)16(19-9-10-20)12-5-3-2-4-6-12/h2-8,11,18,20H,9-10H2,1H3/b19-16-
Formel vægt 288.1029