missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(5-Bromo-2-furyl)thiazolidine, 97%

Brand:  Alfa Aesar™ H62897.06

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15469348

  • 8022.97 DKK / 5g


Estimated Shipment date
from Supplier 20-02-2019
Add to basket

Specifications

Chemical Identifiers

MFCD16685164
5-Bromo-2-(2-thiazolidinyl)furan

Specifications

2-(5-Bromo-2-furyl)thiazolidine
MFCD16685164
234.11
97%
5g
5-Bromo-2-(2-thiazolidinyl)furan
Solid
Videos
SDS