Få mere at vide
2-[(5-Benzyloxypyridin-2-yl)methylamino]ethanol-d4, TRC
Artikelnummer. 30341088
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30341088 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30341088 Leverandør Toronto Research Chemicals Leverandørnr. TRCB2881021MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-[(5-Benzyloxypyridin-2-yl)methylamino]ethanol-d4
CAS 1794786-87-0
Molekylær formel C15H14D4N2O2
Mængde 1 mg
Synonym 2-[Methyl[5-(phenylmethoxy)-2-pyridinyl]amino] Ethanol-d4
InChI formel InChI=1S/C15H18N2O2/c1-17(9-10-18)15-8-7-14(11-16-15)19-12-13-5-3-2-4-6-13/h2-8,11,18H,9-10,12H2,1H3/i9D2,10D2
SMIL [2H]C([2H])(O)C([2H])([2H])N(C)c1ccc(OCc2ccccc2)cn1
IUPAC navn 1,1,2,2-tetradeuterio-2-[methyl-(5-phenylmethoxypyridin-2-yl)amino]ethanol
Molekylvægt (g/mol) 262.34
Formel vægt 262.1619