Få mere at vide
2-(5,5-Dimethyl-1,3,2-dioxaborinan-2-yl)benzoic Acid Ethyl Ester (>90%), TRC
Artikelnummer. 30377753
Skift visning
Mængde:
500 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30377753 500 mg
30378043 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30377753 Leverandør Toronto Research Chemicals Leverandørnr. TRCD472615500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 346656-34-6
Molekylær formel C14 H19 B O4
Molekylvægt (g/mol) 262.11
IUPAC navn ethyl 2-(5,5-dimethyl-1,3,2-dioxaborinan-2-yl)benzoate
SMIL CCOC(=O)c1ccccc1B2OCC(C)(C)CO2
Tekniske data
Kemisk navn eller materiale 2-(5,5-Dimethyl-1,3,2-dioxaborinan-2-yl)benzoic Acid Ethyl E
Mængde 500 mg
InChI formel InChI=1S/C14H19BO4/c1-4-17-13(16)11-7-5-6-8-12(11)15-18-9-14(2,3)10-19-15/h5-8H,4,9-10H2,1-3H3
Formel vægt 262.1376