missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Pyridyl)-7-azabenzimidazole, 97%, Thermo Scientific™

Catalog Number p-7034527
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
63411-78-9
C11H8N4
196.213
MFCD01001152
NEAHTABRXFKZGG-UHFFFAOYSA-N
4-1h-imidazo 4,5-c pyridin-2-yl pyridine, cambridge id 5237049, 2-4-pyridyl-7-azabenzimidazole, 2-4-pyridyl-3h-imidazo 4,5-c pyridine, 2-pyridin-4-yl-3h-imidazo 4,5-c pyridine, 2-4-pyridinyl-1h-imidazo 4,5-c pyridine, 4-3h-imidazo 4,5-c pyridin-2-yl pyridine, 2-pyridin-4-yl-1h-imidazo 4,5-c pyridine, 3h-imidazo 4,5-c pyridine,2-4-pyridinyl
789950
2-pyridin-4-yl-3H-imidazo[4,5-c]pyridine
C1=CN=CC=C1C2=NC3=C(N2)C=NC=C3
This item is not returnable. View return policy
63411-78-9
C11H8N4
196.213
MFCD01001152
NEAHTABRXFKZGG-UHFFFAOYSA-N
4-1h-imidazo 4,5-c pyridin-2-yl pyridine, cambridge id 5237049, 2-4-pyridyl-7-azabenzimidazole, 2-4-pyridyl-3h-imidazo 4,5-c pyridine, 2-pyridin-4-yl-3h-imidazo 4,5-c pyridine, 2-4-pyridinyl-1h-imidazo 4,5-c pyridine, 4-3h-imidazo 4,5-c pyridin-2-yl pyridine, 2-pyridin-4-yl-1h-imidazo 4,5-c pyridine, 3h-imidazo 4,5-c pyridine,2-4-pyridinyl
789950
2-pyridin-4-yl-3H-imidazo[4,5-c]pyridine
C1=CN=CC=C1C2=NC3=C(N2)C=NC=C3

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2-(4-Pyridyl)-7-azabenzimidazole is frequently used as veterinary medicines (mostly anthelmintics). It is also used in pharmaceuticals.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 240°C to 248°C
Quantity 250 mg
Formula Weight 196.21
Percent Purity 97%
Chemical Name or Material 2-(4-Pyridyl)-7-azabenzimidazole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'