missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(4-pyridyl)-1,3-thiazole-4-carboxylic acid, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
10 g
Specifications
CAS Min % | 94.5 |
CAS Max % | 100.0 |
Packaging | Amber glass bottle |
Quantity | 250 mg |
Formula Weight | 206.22 |
Percent Purity | 95% |
Chemical Name or Material | 2-(4-pyridyl)-1, 3-thiazole-4-carboxylic Acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'