Få mere at vide
2,4-Pteridinediamine-6-methanol, TRC
Artikelnummer. 30425552
Skift visning
Mængde:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30425552 100 mg
30410378 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30425552 Leverandør Toronto Research Chemicals Leverandørnr. TRCP840095100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2,4-Pteridinediamine-6-methanol
CAS 945-24-4
Molekylær formel C7 H8 N6 O
Mængde 100 mg
Synonym Methotrexate Ep Impurity A
InChI formel InChI=1S/C7H8N6O/c8-5-4-6(13-7(9)12-5)10-1-3(2-14)11-4/h1,14H,2H2,(H4,8,9,10,12,13)
SMIL Nc1nc(N)c2nc(CO)cnc2n1
IUPAC navn (2,4-diaminopteridin-6-yl)methanol
Molekylvægt (g/mol) 192.18
Formel vægt 192.076