Få mere at vide
2-(4-Nitro-1,3-dioxoisoindolin-2-yl)pentanedioic Acid-d4, TRC
Artikelnummer. 30470859
Skift visning
Mængde:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30470859 25 mg
30479550 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30470859 Leverandør Toronto Research Chemicals Leverandørnr. TRCN51587725MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-(4-Nitro-1,3-dioxoisoindolin-2-yl)pentanedioic Acid-d4
CAS 1416575-82-0
Molekylær formel C13H6D4N2O8
Mængde 25 mg
Synonym 2-(1,3-Dihydro-4-nitro-1,3-dioxo-2H-isoindol-2-yl)pentanedioic Acid-d4, 1-(4-Nitro-1,3-dioxoisoindolin-2-yl)propane-1,3-dicarboxylic Acid-d4
InChI formel InChI=1S/C13H10N2O8/c16-9(17)5-4-8(13(20)21)14-11(18)6-2-1-3-7(15(22)23)10(6)12(14)19/h1-3,8H,4-5H2,(H,16,17)(H,20,21)/i4D2,5D2
SMIL [2H]C([2H])(C(N1C(=O)c2cccc(c2C1=O)[N+](=O)[O-])C(=O)O)C([2H])([2H])C(=O)O
IUPAC navn 2,2,3,3-tetradeuterio-4-(4-nitro-1,3-dioxoisoindol-2-yl)pentanedioic acid
Molekylvægt (g/mol) 326.25
Formel vægt 326.0688